missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(2-Bromoethyl)benzene, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 106941000

 View more versions of this product

Catalog No. AC106941000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(2-Bromoethyl)benzene
103-63-9
100.0
Colorless to Yellow
220°C to 221°C
Authentic
Glass bottle
1.5553 to 1.5573
MFCD00000240
05,356
2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
WMPPDTMATNBGJN-UHFFFAOYSA-N
2-bromoethylbenzene
7666
98%
98%
97.5
-56°C
1.3500g/mL
89°C
97.5% min. (GC)
C8H9Br
C6H5(CH2)2Br
100 mL
1.35
Solubility in water: insoluble. Other solubilities: soluble in ether and benzene
C1=CC=C(C=C1)CCBr
185.06
185.06
Liquid

Chemical Identifiers

103-63-9
WMPPDTMATNBGJN-UHFFFAOYSA-N
7666
C1=CC=C(C=C1)CCBr
MFCD00000240
2-bromoethyl benzene, phenethyl bromide, 2-phenylethyl bromide, 1-bromo-2-phenylethane, phenylethyl bromide, benzene, 2-bromoethyl, 2-phenethyl bromide, beta-bromoethylbenzene, 2-phenyl-1-bromoethane, phenethylbromide
2-bromoethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Warning

EINECSNumber : 203-130-8

RTECSNumber : CY9032000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only