Please login to your online account to display your discounted pricing

(2-Bromoethyl)benzene, 98%, ACROS Organics™


Quantity 2.5L
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC106940025

 View more versions of this product

Catalog No. AC106940025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 185.06
Quantity 2.5L
Color Clear colorless to pale yellow
EINECS Number 203-130-8
TSCA TSCA
Density 1.3500g/mL
RTECS Number CY9032000
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5553 to 1.5573 (20°C, 589nm)
Water 0.05% max.
Assay Percent Range 98%
Synonym β-Bromophenylethane
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Chemical Name or Material (2-Bromoethyl)benzene
Name Note 98%
MDL Number MFCD00000240
Linear Formula C6H5(CH2)2Br
CAS 103-63-9
Molecular Formula C8H9Br
CAS Min % 97.5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material (2-Bromoethyl)benzene
Name Note 98%
CAS 103-63-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9Br
Linear Formula C6H5(CH2)2Br
Formula Weight 185.06
MDL Number MFCD00000240
Synonym β-Bromophenylethane
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.

SDS & Safety Documents