missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoethyl acrylate, 94%, stab. with 0.1% 4-methoxyphenol

Manufacturer:  Alfa Aesar™ L1250203

 View more versions of this product

Catalog No. AAL1250203



Description

Description

Applications
2-Bromoethyl acrylate is utilized in curable and reactive polymers due to the presence of reactive bromine. It is also used in the synthesis of other monomers.

Solubility
Miscible with water.

Notes
Light and heat sensitive Incompatible with strong oxidizing agents, polymerizing agents and peroxides.
Specifications

Specifications

2-Bromoethyl acrylate
1g
4823-47-6
MFCD00013541
2-bromoethyl acrylate, acrylic acid, 2-bromoethyl ester, 2-propenoic acid, 2-bromoethyl ester, ccris 4753, acrylic acid 2-bromoethyl ester, 2-bromoethylacrylate, 2-bromoethyl=acrylate, acmc-20ao1p, acrylic acid 2-bromo-ethyl ester, 2-propenoic acid,2-bromoethyl ester
C=CC(=O)OCCBr
179.013
179.02
52°C to 53°C (5mmHg)
stabilized with 0.1% 4-methoxyphenol
C5H7BrO2
1749713
CDZAAIHWZYWBSS-UHFFFAOYSA-N
2-bromoethyl prop-2-enoate
20963
94%

Chemical Identifiers

4823-47-6
179.013
CDZAAIHWZYWBSS-UHFFFAOYSA-N
20963
C=CC(=O)OCCBr
C5H7BrO2
MFCD00013541
2-bromoethyl acrylate, acrylic acid, 2-bromoethyl ester, 2-propenoic acid, 2-bromoethyl ester, ccris 4753, acrylic acid 2-bromoethyl ester, 2-bromoethylacrylate, 2-bromoethyl=acrylate, acmc-20ao1p, acrylic acid 2-bromo-ethyl ester, 2-propenoic acid,2-bromoethyl ester
2-bromoethyl prop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AS4810000

Recommended Storage : 2° to 8°C

Documents