Learn More
2-Bromoethanol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A102750B
Description
2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromoethanol is utilized in selective reduction of nitroarenes. It is used as a solvent and organic synthetic raw material. It finds application in the synthesis of 2-piperidin-1-yl-ethanol by reacting with piperidine. It is used in selective reduction of nitroarenes.
Solubility
Miscible with water, organic solvent and petroleum ether.
Notes
Light sensitive. Hygroscopic. Store in a cool place. Incompatible with strong acids, strong oxidizing agents, strong reducing agents, acid chlorides and acid anhydrides.
Specifications
2-Bromoethanol | |
-80°C | |
149°C to 150°C | |
C2H5BrO | |
BrCH2CH2OH | |
1000 g | |
878140 | |
14,3792 | |
Miscible with water,organic solvent and petroleum ether. | |
C(CBr)O | |
124.965 | |
124.97 |
540-51-2 | |
1.762 | |
>110°C (230°F) | |
1.493 | |
MFCD00002827 | |
UN2922 | |
Hygroscopic; Light sensitive | |
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol | |
LDLCZOVUSADOIV-UHFFFAOYSA-N | |
2-bromoethanol | |
10898 | |
97% |
Chemical Identifiers
540-51-2 | |
124.965 | |
LDLCZOVUSADOIV-UHFFFAOYSA-N | |
10898 | |
C(CBr)O |
C2H5BrO | |
MFCD00002827 | |
ethylene bromohydrin, ethanol, 2-bromo, 2-bromoethan-1-ol, glycol bromohydrin, 2-hydroxyethyl bromide, 2-bromo-1-ethanol, 1-bromo-2-ethanol, ethylenebromohydrin, 2-bromoethyl alcohol, 2-bromo-ethanol | |
2-bromoethanol |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P262-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H331-H310-H314
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 208-748-1
RTECSNumber : KJ8225000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only