missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromodibenzothiophene 5,5-Dioxide 95.0+%, TCI America™
$227.29 - $678.52
Chemical Identifiers
CAS | 53846-85-8 |
---|---|
Molecular Formula | C12H7BrO2S |
Molecular Weight (g/mol) | 295.15 |
InChI Key | WRKJLRFOQLWYOW-UHFFFAOYSA-N |
Synonym | 2-Bromodibenzothiophene Sulfone, 2-Bromodibenzosulfolane |
PubChem CID | 554821 |
IUPAC Name | 2-bromodibenzothiophene 5,5-dioxide |
SMILES | C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=CC(=C3)Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B5267200MG
|
TCI America
B5267200MG |
200 mg |
Each for $227.29
|
|
|||||
B52671G
|
TCI America
B52671G |
1 g |
Each for $678.52
|
|
|||||
Chemical Identifiers
53846-85-8 | |
295.15 | |
2-Bromodibenzothiophene Sulfone, 2-Bromodibenzosulfolane | |
2-bromodibenzothiophene 5,5-dioxide |
C12H7BrO2S | |
WRKJLRFOQLWYOW-UHFFFAOYSA-N | |
554821 | |
C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=CC(=C3)Br |
Specifications
53846-85-8 | |
C12H7BrO2S | |
2-Bromodibenzothiophene Sulfone, 2-Bromodibenzosulfolane | |
C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=CC(=C3)Br | |
295.15 | |
295.15 | |
Crystalline Powder |
267°C | |
200 mg | |
WRKJLRFOQLWYOW-UHFFFAOYSA-N | |
2-bromodibenzothiophene 5,5-dioxide | |
554821 | |
≥95.0% (GC) | |
2-Bromodibenzothiophene 5,5-Dioxide |
Safety and Handling
TSCA : No