missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromodibenzothiophene 5,5-Dioxide 95.0+%, TCI America™
Supplier: TCI America B52671G
Specifications
2-Bromodibenzothiophene 5,5-Dioxide | |
267°C | |
1 g | |
WRKJLRFOQLWYOW-UHFFFAOYSA-N | |
2-bromodibenzothiophene 5,5-dioxide | |
554821 | |
≥95.0% (GC) |
53846-85-8 | |
C12H7BrO2S | |
2-Bromodibenzothiophene Sulfone, 2-Bromodibenzosulfolane | |
C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=CC(=C3)Br | |
295.15 | |
295.15 | |
Crystalline Powder |
Chemical Identifiers
53846-85-8 | |
295.15 | |
2-Bromodibenzothiophene Sulfone, 2-Bromodibenzosulfolane | |
2-bromodibenzothiophene 5,5-dioxide |
C12H7BrO2S | |
WRKJLRFOQLWYOW-UHFFFAOYSA-N | |
554821 | |
C1=CC=C2C(=C1)C3=C(S2(=O)=O)C=CC(=C3)Br |
Safety and Handling
TSCA : No