Please login to your online account to display your discounted pricing

2-Bromobutane, 99+%, ACROS Organics™


Quantity 100mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  106781000

 View more versions of this product

Catalog No. AC106781000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Formula Weight 137.02
Physical Form Liquid
Quantity 100mL
Color Clear colorless to yellow-brown
RTECS Number EJ6228000
EINECS Number 201-140-7
TSCA TSCA
CAS Max % 100.0
CAS Min % 99.0
Synonym sec-Butyl bromide
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Chemical Name or Material 2-Bromobutane
Linear Formula C2H5CH(Br)CH3
Molecular Formula C4H9Br
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
MDL Number MFCD00000156
Assay Percent Range 99+%
Name Note 99+%
CAS 78-76-2
Refractive Index 1.4359 to 1.4379 (20°C, 589nm)
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-Bromobutane
Name Note 99+%
CAS 78-76-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C4H9Br
Linear Formula C2H5CH(Br)CH3
Formula Weight 137.02
MDL Number MFCD00000156
Synonym sec-Butyl bromide
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.

SDS & Safety Documents