Learn More
2-Bromobutane, 99+%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 106780025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Bromobutane | |
78-76-2 | |
100.0 | |
91°C | |
Authentic | |
Glass bottle | |
1.4359 to 1.4379 | |
MFCD00000156 | |
01,119 | |
15,1556 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether | |
CCC(C)Br | |
137.02 | |
137.02 |
99+% | |
99.0 | |
1.2500g/mL | |
21°C | |
99% min. (GC) | |
C4H9Br | |
C2H5CH(Br)CH3 | |
2.5L | |
1.25 | |
sec-butyl bromide, butane, 2-bromo, 2-butyl bromide, methylethylbromomethane, 2-bromo-butane, secondary butyl bromide, sec-butylbromide, 1-bromo-1-methylpropane, ccris 106, bromobutane, 2 | |
UPSXAPQYNGXVBF-UHFFFAOYSA-N | |
2-bromobutane | |
6554 | |
99+% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a
GHS Signal Word: Danger
EINECSNumber : 201-140-7
RTECSNumber : EJ6228000
TSCA : TSCA
RUO â Research Use Only