missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromobenzylsulfonyl chloride, 97%, Maybridge

Chemical Identifiers

CAS 24974-74-1
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C7 H6 Br Cl O2 S
Formula Weight 269.55
MDL Number MFCD06798044
Chemical Name or Material 2-Bromobenzylsulfonyl chloride
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MO07945DE SDS Fisher Scientific Company
MO07945DE
Amber Glass Bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

24974-74-1
97%
269.55
2-Bromobenzylsulfonyl chloride
97.0
C7 H6 Br Cl O2 S
MFCD06798044
97%

Specifications

269.55
97.0
24974-74-1
MFCD06798044
2-Bromobenzylsulfonyl chloride
5g
Amber Glass Bottle
C7 H6 Br Cl O2 S
97%
97%

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger