Learn More
2-Bromobenzylamine hydrochloride, 97%, ACROS Organics™
$421.11 - $421.11
Chemical Identifiers
CAS | 5465-63-4 |
---|---|
Molecular Formula | C7H8BrN·HCl |
Molecular Weight (g/mol) | 222.52 |
MDL Number | MFCD00012853 |
InChI Key | MDEJOHXOXKDGOU-UHFFFAOYSA-N |
Synonym | 1-2-bromophenyl methanamine hydrochloride, 1-2-bromophenyl methylamine hydrochloride, 2-bromobenzylamine hcl, 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, 2-bromophenyl methylamine, chloride, acmc-1aup2, benzenemethanamine, 2-bromo-, hydrochloride, o-bromobenzylamine hydrochloride, pubchem3749 |
PubChem CID | 2724202 |
IUPAC Name | (2-bromophenyl)methanamine;hydrochloride |
SMILES | C1=CC=C(C(=C1)CN)Br.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC209090250
|
Thermo Fisher Scientific
209090250 |
25g | Glass bottle |
Each for $421.11
|
|
Chemical Identifiers
5465-63-4 | |
222.52 | |
MDEJOHXOXKDGOU-UHFFFAOYSA-N | |
2724202 | |
C1=CC=C(C(=C1)CN)Br.Cl |
C7H8BrN·HCl | |
MFCD00012853 | |
1-2-bromophenyl methanamine hydrochloride, 1-2-bromophenyl methylamine hydrochloride, 2-bromobenzylamine hcl, 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, 2-bromophenyl methylamine, chloride, acmc-1aup2, benzenemethanamine, 2-bromo-, hydrochloride, o-bromobenzylamine hydrochloride, pubchem3749 | |
(2-bromophenyl)methanamine;hydrochloride |
Specifications
Authentic | |
0.5% max. (K.F.) | |
225°C to 230°C | |
5465-63-4 | |
100.0 | |
C7H8BrN·HCl | |
MFCD00012853 | |
1-2-bromophenyl methanamine hydrochloride, 1-2-bromophenyl methylamine hydrochloride, 2-bromobenzylamine hcl, 2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, 2-bromophenyl methylamine, chloride, acmc-1aup2, benzenemethanamine, 2-bromo-, hydrochloride, o-bromobenzylamine hydrochloride, pubchem3749 | |
C1=CC=C(C(=C1)CN)Br.Cl | |
222.52 | |
222.52 | |
97% |
Glass bottle | |
White to Yellow | |
25g | |
96.0 | |
96% min. (Argentometry) | |
BrC6H4CH2NH2·HCl | |
12, 1074 | |
MDEJOHXOXKDGOU-UHFFFAOYSA-N | |
(2-bromophenyl)methanamine;hydrochloride | |
2724202 | |
Crystalline Powder, Crystals and/or Chunks | |
2-Bromobenzylamine hydrochloride, 97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
TSCA : TSCA