missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromobenzylamine, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC350510010

 View more versions of this product

Catalog No. AC350510010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Bromobenzylamine
752160
97.5
Glass bottle
Clear
Liquid
98%
BrC6H4CHNH
MFCD00025572
98%
Authentic
100.0
1.5875 to 1.5915 (20°C, 589nm)
1g
≥97.5% (GC)
C7H8BrN
186.05

Chemical Identifiers

752160
100.0
C7H8BrN
186.05
2-Bromobenzylamine, 98%
97.5
98%
BrC6H4CHNH
MFCD00025572
≥97.5% (GC)

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger