missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromobenzyl alcohol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1570009
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromobenzyl alcohol | |
78°C to 82°C | |
MFCD00004600 | |
2243418 | |
IOWGHQGLUMEZKG-UHFFFAOYSA-N | |
(2-bromophenyl)methanol | |
72850 | |
98% |
18982-54-2 | |
C7H7BrO | |
10 g | |
2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746 | |
OCC1=CC=CC=C1Br | |
187.04 | |
187.04 |
Chemical Identifiers
18982-54-2 | |
187.04 | |
IOWGHQGLUMEZKG-UHFFFAOYSA-N | |
72850 | |
OCC1=CC=CC=C1Br |
C7H7BrO | |
MFCD00004600 | |
2-bromobenzyl alcohol, 2-bromophenyl methanol, o-bromobenzyl alcohol, benzenemethanol, 2-bromo, o-bromo-benzyl alcohol, 2-bromobenzylalcohol, 2-bromophenyl methan-1-ol, o-bromobenzylalcohol, 2-bromo-benzylalcohol, pubchem3746 | |
(2-bromophenyl)methanol |
Safety and Handling
P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c
H302+H332-H319
EINECSNumber : 242-719-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only