missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromobenzoyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1452018

 View more versions of this product

Catalog No. AAA1452018



Specifications

Specifications

2-Bromobenzoyl chloride
245°C
50g
C7H4BrClO
UN3265
benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
C1=CC=C(C(=C1)C(=O)Cl)Br
219.462
219.47
107°C (224°F)
10°C
7154-66-7
MFCD00000655
508506
NZCKTGCKFJDGFD-UHFFFAOYSA-N
2-bromobenzoyl chloride
23542
98%

Chemical Identifiers

7154-66-7
219.462
NZCKTGCKFJDGFD-UHFFFAOYSA-N
23542
C1=CC=C(C(=C1)C(=O)Cl)Br
C7H4BrClO
MFCD00000655
benzoyl chloride, 2-bromo, o-bromobenzoyl chloride, benzoyl chloride, o-bromo, 2-bromo-benzoyl chloride, 2-bromobenzoylchloride, 2-bromobenzene-1-carbonyl chloride, o-brombenzoylchlorid, bromo benzoylchloride, o-bromobenzoylchloride, pubchem12510
2-bromobenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : DM6635000

Documents