Please login to your online account to display your discounted pricing

2-Bromobenzotrifluoride, 99%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC106735000

 View more versions of this product

Catalog No. AC106735000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 2-Bromobenzotrifluoride
CAS 392-83-6
Infrared Spectrum Authentic
CAS Min % 98.5
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4810 to 1.4830 (20°C, 589nm)
Density 1.65g/mL
Color Clear colorless to brown
Quantity 500g
Physical Form Liquid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4BrF3
Linear Formula BrC6H4CF3
Formula Weight 225.01
MDL Number MFCD00000373
Synonym o-Bromobenzotrifluoride

Specifications

Chemical Name or Material 2-Bromobenzotrifluoride
CAS 392-83-6
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C7H4BrF3
Linear Formula BrC6H4CF3
Formula Weight 225.01
MDL Number MFCD00000373
Synonym o-Bromobenzotrifluoride
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents