missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromobenzothiazole, 99%

Manufacturer:  Alfa Aesar™ H6328614

 View more versions of this product

Catalog No. AAH6328614



Description

Description

Applications
2-Bromobenzothiazole, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, acids and bases.
Specifications

Specifications

2-Bromobenzothiazole
39°C to 43°C
2516-40-7
MFCD02681887
DRLMMVPCYXFPEP-UHFFFAOYSA-N
2-bromo-1,3-benzothiazole
612040
Crystalline Solid
Yellow
25g
C7H4BrNS
2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
C1=CC=C2C(=C1)N=C(S2)Br
214.08
214.08
99%

Chemical Identifiers

2516-40-7
214.08
DRLMMVPCYXFPEP-UHFFFAOYSA-N
612040
C1=CC=C2C(=C1)N=C(S2)Br
C7H4BrNS
MFCD02681887
2-bromobenzothiazole, 2-bromobenzo d thiazole, benzothiazole, 2-bromo, dsstox_cid_1498, dsstox_rid_76188, dsstox_gsid_21498, 2-bromobenzthiazole, 2-bromo benzothiazole, 2-bromo-benzothiazole, acmc-20a0ou
2-bromo-1,3-benzothiazole
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents