Learn More
2-Bromobenzimidazole, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2618306
Description
Reactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReactant in the synthesis of substituted benzimidazoles. 2-Bromobenzimidazole is the starting compound in the synthesis of polyhalogenobenzimidazoles.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
2-Bromobenzimidazole | |
195°C to 201°C | |
MFCD00160009 | |
2-bromobenzimidazole, 2-bromo-1h-benzo d imidazole, 2-bromo-1h-1,3-benzodiazole, 2-bromo-1h-benzoimidazole, 1h-benzimidazole, 2-bromo, bromobenzimidazole, bromo-benzimidazole, pubchem16113, acmc-1an7o, 1h-benzimidazole,2-bromo | |
PHPYXVIHDRDPDI-UHFFFAOYSA-N | |
2-bromo-1H-benzimidazole | |
2776281 | |
99% |
54624-57-6 | |
C7H5BrN2 | |
5 g | |
Insoluble in water. | |
C1=CC=C2C(=C1)NC(=N2)Br | |
197.035 | |
197.03 |
Chemical Identifiers
54624-57-6 | |
197.035 | |
PHPYXVIHDRDPDI-UHFFFAOYSA-N | |
2-bromo-1H-benzimidazole |
C7H5BrN2 | |
MFCD00160009 | |
2776281 | |
C1=CC=C2C(=C1)NC(=N2)Br |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
RUO – Research Use Only