missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromobenzenesulfonyl chloride, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 306440050

 View more versions of this product

Catalog No. AC306440050


 

Specifications Description & Specifications

Specifications

2-Bromobenzenesulfonyl chloride
2905-25-1
96.0
Glass bottle
48° to 52°C
Crystalline Powder
97%
BrC6H4SOCl
MFCD00051971
97%
Authentic
100.0
Slightly yellow
5g
≥96.0%
C6H4BrClO2S
255.52

Chemical Identifiers

2905-25-1
100.0
C6H4BrClO2S
255.52
2-Bromobenzenesulfonyl chloride, 97%
96.0
97%
BrC6H4SOCl
MFCD00051971
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA