Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromobenzenesulfonamide, 99%

$53.57 - $167.78

Chemical Identifiers

Chemical Name or Material 2-Bromobenzenesulfonamide
CAS 92748-09-9
Molecular Formula C6H6BrNO2S
Formula Weight 236.09
MDL Number MFCD00173662
Beilstein 3253041
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1757803

 
alfa aesar™
L1757803
1g Each for $53.57

AAL1757806

 
alfa aesar™
L1757806
5g Each for $167.78
Description & Specifications

Specifications

Formula Weight 236.09
Chemical Name or Material 2-Bromobenzenesulfonamide
Assay 99%
Molecular Formula C6H6BrNO2S
MDL Number MFCD00173662
CAS 92748-09-9
Beilstein 3253041
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 191° to 192°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.