Please login to your online account to display your discounted pricing

2-Bromobenzamide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Bromobenzamide
CAS 4001-73-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H6BrNO
Linear Formula BrC6H4C(=O)NH2
Formula Weight 200.03
MDL Number MFCD00007969
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183990050

  SDS
thermo fisher scientific
AC183990050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 200.03
Quantity 5g
Color Beige
Melting Point 158° to 162°C
Beilstein 09, 348
CAS Max % 100.0
Chemical Name or Material 2-Bromobenzamide
Assay Percent Range 98%
Molecular Formula C7H6BrNO
Linear Formula BrC6H4C(=O)NH2
MDL Number MFCD00007969
CAS 4001-73-4
Infrared Spectrum Authentic
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.