Please login to your online account to display your discounted pricing

2-Bromobenzamide, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  RDP00161EA

 View more versions of this product

Catalog No. RDP00161EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 10g
Assay 97%
Formula Weight 200.03
Chemical Name or Material 2-Bromobenzamide
Assay Percent Range 97%
CAS 4001-73-4
Linear Formula BrC6H4C(=O)NH2
Packaging Amber Glass Bottle
Molecular Formula C7 H6 Br N O

Specifications

Chemical Name or Material 2-Bromobenzamide
Assay 97%
Assay Percent Range 97%
Molecular Formula C7 H6 Br N O
Linear Formula BrC6H4C(=O)NH2
Formula Weight 200.03