Please login to your online account to display your discounted pricing

2-Bromobenzamide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Bromobenzamide
Assay 97%
Assay Percent Range 97%
Molecular Formula C7 H6 Br N O
Linear Formula BrC6H4C(=O)NH2
Formula Weight 200.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RDP00161EA

  SDS
fisher scientific company
RDP00161EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10g
Assay 97%
Formula Weight 200.03
Chemical Name or Material 2-Bromobenzamide
Assay Percent Range 97%
CAS 4001-73-4
Linear Formula BrC6H4C(=O)NH2
Packaging Amber Glass Bottle
Molecular Formula C7 H6 Br N O