Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromobenzaldoxime, 96%

$132.53 - $132.53

Chemical Identifiers

Chemical Name or Material 2-Bromobenzaldoxime
CAS 34158-72-0
Molecular Formula C7H6BrNO
Formula Weight 200.03
MDL Number MFCD00082734
Beilstein 471599
Synonym 2-Bromobenzaldehyde oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2302306

  SDS
alfa aesar™
B2302306
5g Each for $132.53
Description & Specifications

Specifications

Formula Weight 200.03
Chemical Name or Material 2-Bromobenzaldoxime
Melting Point 100° to 104°C
Quantity 5g
Assay 96%
Molecular Formula C7H6BrNO
MDL Number MFCD00082734
CAS 34158-72-0
Beilstein 471599
Synonym 2-Bromobenzaldehyde oxime
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.