Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromobenzaldehyde ethylene acetal, 98+%

$44.45 - $152.19

Chemical Identifiers

Chemical Name or Material 2-Bromobenzaldehyde ethylene acetal
CAS 34824-58-3
Molecular Formula C9H9BrO2
Formula Weight 229.07
MDL Number MFCD00155124
Beilstein 1284374
Synonym 2-(2-Bromophenyl)-1,3-dioxolane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1960406

  SDS
alfa aesar™
L1960406
5g Each for $44.45

AAL1960414

  SDS
alfa aesar™
L1960414
25g Each for $152.19
Description & Specifications

Specifications

Formula Weight 229.07
Chemical Name or Material 2-Bromobenzaldehyde ethylene acetal
Molecular Formula C9H9BrO2
Assay ≥98%
MDL Number MFCD00155124
Synonym 2-(2-Bromophenyl)-1,3-dioxolane
CAS 34824-58-3
Boiling Point 126° to 127°C (5mmHg)
Quantity 5g
Beilstein 1284374

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed. Incompatible with oxidizing agents.