Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromobenzaldehyde diethyl acetal, 98%

$25.47 - $142.24

Chemical Identifiers

Chemical Name or Material 2-Bromobenzaldehyde diethyl acetal
CAS 35822-58-3
Molecular Formula C11H15BrO2
Formula Weight 259.14
MDL Number MFCD01075696
Beilstein 5935613
Synonym 2-Bromo-alpha,alpha-diethoxytoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1551306

  SDS
alfa aesar™
L1551306
5g Each for $25.47

AAL1551314

  SDS
alfa aesar™
L1551314
25g Each for $64.65

AAL1551322

  SDS
alfa aesar™
L1551322
100g Each for $142.24
Description & Specifications

Specifications

Formula Weight 259.14
Chemical Name or Material 2-Bromobenzaldehyde diethyl acetal
Assay 98%
Molecular Formula C11H15BrO2
MDL Number MFCD01075696
CAS 35822-58-3
Beilstein 5935613
Synonym 2-Bromo-alpha,alpha-diethoxytoluene
Boiling Point 95°C (1mmHg)
Flash Point 113°C (235°F)
Quantity 5g