missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoanisole, 97%, ACROS Organics™

Manufacturer:  Acros Organics 170261000

 View more versions of this product

Catalog No. AC170261000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Bromoanisole
Glass bottle
Colorless to Yellow
97%
96.0
97%
BrC6H4OCH3
06, 197
HTDQSWDEWGSAMN-UHFFFAOYSA-N
1-bromo-2-methoxybenzene
11358
Liquid
Authentic
1.5730 to 1.5750 (20°C, 589nm)
100mL
578-57-4
100.0
C7H7BrO
MFCD00000064
2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
COC1=CC=CC=C1Br
187.036
187.04
≥96.0%

Chemical Identifiers

578-57-4
187.036
HTDQSWDEWGSAMN-UHFFFAOYSA-N
11358
COC1=CC=CC=C1Br
C7H7BrO
MFCD00000064
2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
1-bromo-2-methoxybenzene
Safety and Handling

Safety and Handling

EINECSNumber : 209-425-8

RTECSNumber : BZ8500000

TSCA : TSCA

SDS Documents