missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoanisole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 170260250

 View more versions of this product

Catalog No. AC170260250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Bromoanisole
578-57-4
100.0
223°C
Authentic
Glass bottle
1.5730 to 1.575
MFCD00000064
06, 197
2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
HTDQSWDEWGSAMN-UHFFFAOYSA-N
1-bromo-2-methoxybenzene
11358
97%
97%
96.0
1.5000g/mL
96°C
96% min. (GC)
C7H7BrO
BrC6H4OCH3
25mL
1.5
Solubility in water: immiscible
COC1=CC=CC=C1Br
187.04
187.04

Chemical Identifiers

578-57-4
187.04
HTDQSWDEWGSAMN-UHFFFAOYSA-N
11358
COC1=CC=CC=C1Br
C7H7BrO
MFCD00000064
2-bromoanisole, o-bromoanisole, benzene, 1-bromo-2-methoxy, 2-methoxybromobenzene, o-anisyl bromide, anisole, o-bromo, o-methoxybromobenzene, 2-bromoanisol, o-methoxyphenyl bromide, 2-methoxyphenyl bromide
1-bromo-2-methoxybenzene
Safety and Handling

Safety and Handling

EINECSNumber : 209-425-8

RTECSNumber : BZ8500000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only