missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromoaniline, 98%

Manufacturer:  Alfa Aesar™ A1178836

 View more versions of this product

Catalog No. AAA1178836


 

Description

Description

Applications
2-Bromoaniline is used as a corrosion inhibitor, emulsifying and antiseptic agents. It is used in the rubber industry, e.g. diphenylguanidines, phenylenediamines mercaptobenzothiazoles, aniline ketones and etc.

Solubility
Insoluble in water.

Notes
Store at 2-8°C. Incompatible with acids, Acid chlorides, Acid anhydrides, Chloroformates, Strong oxidizing agents.
Specifications

Specifications

2-Bromoaniline
98%
MFCD00007632
27°C to 31°C
500g
Characteristic
C1=CC=C(C(=C1)N)Br
172.025
172.03
615-36-1
C6H6BrN
UN2811
228°C to 230°C
>110°C (230°F)
AOPBDRUWRLBSDB-UHFFFAOYSA-N
2-bromoaniline
11992

Chemical Identifiers

615-36-1
172.025
AOPBDRUWRLBSDB-UHFFFAOYSA-N
C6H6BrN
MFCD00007632
11992
Safety and Handling

Safety and Handling

GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310-P302+P352
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

SDS Documents