missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Bromoacetyl)-6-methoxynaphthalene, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC450060010
Specifications
2-(Bromoacetyl)-6-methoxynaphthalene | |
112.0°C | |
10262-65-4 | |
C13H11BrO2 | |
Solubility in water: insoluble | |
COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr | |
279.13 | |
279.13 |
Glass Bottle | |
1g | |
95% min. (HPLC) | |
2-bromo-1-6-methoxy-2-naphthyl ethanone, 2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone, 2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one, 2-bromo-1-6-methoxynaphthalen-2-yl ethanone, 2-bromo-6'-methoxy-2'-acetonaphthone, 2-bromoacetyl-6-methoxynaphthalene, enamine_005243, ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl | |
HHHKEQGAGUAOQI-UHFFFAOYSA-N | |
2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone | |
193452 | |
96% |
Chemical Identifiers
10262-65-4 | |
279.13 | |
2-bromo-1-6-methoxy-2-naphthyl ethanone, 2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone, 2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one, 2-bromo-1-6-methoxynaphthalen-2-yl ethanone, 2-bromo-6'-methoxy-2'-acetonaphthone, 2-bromoacetyl-6-methoxynaphthalene, enamine_005243, ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl | |
2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone |
C13H11BrO2 | |
HHHKEQGAGUAOQI-UHFFFAOYSA-N | |
193452 | |
COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr |
Safety and Handling
GHS Signal Word: Danger