missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Bromoacetophenone, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1198218

 View more versions of this product

Catalog No. AAA1198218



Description

Description

2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
Specifications

Specifications

2'-Bromoacetophenone
1.476
>110°C (230°F)
1.568
50 g
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
PIMNFNXBTGPCIL-UHFFFAOYSA-N
1-(2-bromophenyl)ethanone
75060
99%
2142-69-0
116°C to 117°C (12 mmHg)
C8H7BrO
MFCD00000067
1931534
Insoluble in water.
CC(=O)C1=CC=CC=C1Br
199.05
199.05

Chemical Identifiers

2142-69-0
199.05
PIMNFNXBTGPCIL-UHFFFAOYSA-N
75060
CC(=O)C1=CC=CC=C1Br
C8H7BrO
MFCD00000067
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
1-(2-bromophenyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 218-398-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only