Learn More
2'-Bromoacetophenone, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1198230
Description
2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-bromoacetophenone is employed in the selective derivatization of cytosine moieties for the determination of global DNA methylation by reversed phase high performance liquid chromatography with spectrofluorimetric detection.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
2'-Bromoacetophenone | |
1.476 | |
>110°C (230°F) | |
1.568 | |
250 g | |
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone | |
PIMNFNXBTGPCIL-UHFFFAOYSA-N | |
1-(2-bromophenyl)ethanone | |
75060 | |
99% |
2142-69-0 | |
116°C to 117°C (12 mmHg) | |
C8H7BrO | |
MFCD00000067 | |
1931534 | |
Insoluble in water. | |
CC(=O)C1=CC=CC=C1Br | |
199.05 | |
199.05 |
Chemical Identifiers
2142-69-0 | |
199.05 | |
PIMNFNXBTGPCIL-UHFFFAOYSA-N | |
75060 | |
CC(=O)C1=CC=CC=C1Br |
C8H7BrO | |
MFCD00000067 | |
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone | |
1-(2-bromophenyl)ethanone |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 218-398-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only