Learn More
2-Bromoacetophenone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 152015000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagentSpecifications
2-Bromoacetophenone | |
70-11-1 | |
100.0 | |
Brown-Green to White | |
>112°C | |
97.5% min. (GC) | |
C8H7BrO | |
MFCD00000195 | |
07, 283 | |
15, 1409 | |
Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chloroform,and ether | |
C1=CC=C(C=C1)C(=O)CBr | |
199.05 | |
CHEBI:51846 | |
98% |
98% | |
97.5 | |
46.0°C to 51.0°C | |
135.0°C (18.0 mmHg) | |
Authentic | |
Plastic bottle | |
C6H5COCH2Br | |
500 g | |
02,316; 12,381 | |
2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo | |
LIGACIXOYTUXAW-UHFFFAOYSA-N | |
2-bromo-1-phenylethanone | |
6259 | |
199.05 | |
Crystals or Powder |
Chemical Identifiers
70-11-1 | |
199.05 | |
LIGACIXOYTUXAW-UHFFFAOYSA-N | |
6259 | |
2-bromo-1-phenylethanone |
C8H7BrO | |
MFCD00000195 | |
2-bromoacetophenone, phenacyl bromide, 2-bromo-1-phenylethan-1-one, bromoacetophenone, ethanone, 2-bromo-1-phenyl, bromomethyl phenyl ketone, alpha-bromoacetophenone, stauffer 4644, omega-bromoacetophenone, acetophenone, 2-bromo | |
CHEBI:51846 | |
C1=CC=C(C=C1)C(=O)CBr |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 200-724-9
TSCA : TSCA
RUO – Research Use Only