Please login to your online account to display your discounted pricing

Alfa Aesar™ 2'-Bromoacetanilide, 98+%

$68.81 - $217.04

Chemical Identifiers

Chemical Name or Material 2'-Bromoacetanilide
CAS 614-76-6
Molecular Formula C8H8BrNO
Formula Weight 214.06
MDL Number MFCD00099252
Beilstein 2086970
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2013206

  SDS
alfa aesar™
L2013206
5g Each for $68.81

AAL2013214

  SDS
alfa aesar™
L2013214
25g Each for $217.04
Description & Specifications

Specifications

Formula Weight 214.06
Chemical Name or Material 2'-Bromoacetanilide
Molecular Formula C8H8BrNO
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00099252
CAS 614-76-6
Assay ≥98%
Melting Point 99°C
Beilstein 2086970
Quantity 5g
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.