missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Bromoacetanilide, 96%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC436330250
Specifications
2'-Bromoacetanilide | |
White | |
25g | |
95.0 | |
95% min. (GC) | |
MFCD00099252 | |
VOBKUOHHOWQHFZ-UHFFFAOYSA-N | |
N-(2-bromophenyl)acetamide | |
136416 | |
Crystalline Powder |
Authentic | |
100°C to 102°C | |
614-76-6 | |
100.0 | |
C8H8BrNO | |
2'-bromoacetanilide, 2-bromo-n-acetylaniline, 2/'-bromoacetanilide, acetamide, n-2-bromophenyl, acetanilide, 2'-bromo, acmc-209msx, n-2-bromo-phenyl-acetamide, n-2-bromophenyl acetamide, o-bromoacetanilide, ortho-bromoacetanilide | |
CC(=O)NC1=CC=CC=C1Br | |
214.06 | |
214.06 | |
96% |
Chemical Identifiers
614-76-6 | |
214.06 | |
VOBKUOHHOWQHFZ-UHFFFAOYSA-N | |
136416 | |
CC(=O)NC1=CC=CC=C1Br |
C8H8BrNO | |
MFCD00099252 | |
2'-bromoacetanilide, 2-bromo-n-acetylaniline, 2/'-bromoacetanilide, acetamide, n-2-bromophenyl, acetanilide, 2'-bromo, acmc-209msx, n-2-bromo-phenyl-acetamide, n-2-bromophenyl acetamide, o-bromoacetanilide, ortho-bromoacetanilide | |
N-(2-bromophenyl)acetamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning