missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoacetamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 291100250

 View more versions of this product

Catalog No. AC291100250



Description

Description

Specifications

Specifications

2-Bromoacetamide
Glass bottle
88°C to 91°C
98%
97.5
97.5% min. (GC)
BrCH2CONH2
02, 216
JUIKUQOUMZUFQT-UHFFFAOYSA-N
2-bromoacetamide
69632
Crystalline Powder
Authentic
White
25g
683-57-8
100.0
C2H4BrNO
MFCD00008025
2-bromo acetamide, 2-bromo-acetamide, 2-bromoacetoamide, acetamide, 2-bromo, alpha-bromoacetamide, bromacetamide, bromo acetamide, bromoacetamide, bromoacetoamide, sjyhcabiktp@
C(C(=O)N)Br
137.96
137.96
98%

Chemical Identifiers

683-57-8
137.96
JUIKUQOUMZUFQT-UHFFFAOYSA-N
69632
C(C(=O)N)Br
C2H4BrNO
MFCD00008025
2-bromo acetamide, 2-bromo-acetamide, 2-bromoacetoamide, acetamide, 2-bromo, alpha-bromoacetamide, bromacetamide, bromo acetamide, bromoacetamide, bromoacetoamide, sjyhcabiktp@
2-bromoacetamide
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

RTECSNumber : AB4587000

TSCA : TSCA

SDS Documents
Product Certifications