missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromoacetamide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 291100050

 View more versions of this product

Catalog No. AC291100050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromoacetamide
683-57-8
100.0
White
97.5% min. (GC)
C2H4BrNO
MFCD00008025
02, 216
JUIKUQOUMZUFQT-UHFFFAOYSA-N
2-bromoacetamide
69632
98%
98%
97.5
88°C to 91°C
Authentic
Glass bottle
BrCH2CONH2
5 g
bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
C(C(=O)N)Br
137.96
137.96
Crystalline Powder

Chemical Identifiers

683-57-8
137.96
JUIKUQOUMZUFQT-UHFFFAOYSA-N
69632
C(C(=O)N)Br
C2H4BrNO
MFCD00008025
bromoacetamide, acetamide, 2-bromo, 2-bromo-acetamide, alpha-bromoacetamide, bromacetamide, bromoacetoamide, bromo acetamide, 2-bromoacetoamide, 2-bromo acetamide, sjyhcabiktp@
2-bromoacetamide
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Do not breat

GHS Signal Word: Danger

RTECSNumber : AB4587000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only