Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-p-xylene, 98+%

$80.96 - $213.96

Chemical Identifiers

Chemical Name or Material 2-Bromo-p-xylene
CAS 553-94-6
Molecular Formula C8H9Br
Formula Weight 185.07
MDL Number MFCD00000074
Beilstein 2040350
Synonym 2-Bromo-1,4-dimethylbenzene; 2,5-Dimethylbromobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1435822

 
alfa aesar™
A1435822
100g Each for $213.96

AAA1435814

 
alfa aesar™
A1435814
25g Each for $80.96
Description & Specifications

Specifications

Formula Weight 185.07
Chemical Name or Material 2-Bromo-p-xylene
TSCA TSCA
Melting Point 9° to 10°C
Boiling Point 199° to 201°C
Quantity 100g
Flash Point 79°C (174°F)
Assay ≥98%
Molecular Formula C8H9Br
MDL Number MFCD00000074
CAS 553-94-6
Beilstein 2040350
Synonym 2-Bromo-1,4-dimethylbenzene; 2,5-Dimethylbromobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.