Learn More
2-Bromo-p-xylene, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 173420250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-p-xylene | |
97% | |
Colorless | |
100.0 | |
199°C to 201°C | |
Authentic | |
Liquid | |
96% min. (GC) | |
C8H9Br | |
MFCD00000074 | |
2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo | |
CC1=CC=C(C)C(Br)=C1 | |
185.06 | |
185.06 |
9°C to 10°C | |
553-94-6 | |
96.0 | |
1.3400g/mL | |
25 g | |
Glass bottle | |
1.5495 to 1.5515 | |
1.34 | |
(CH3)2C6H3Br | |
05, 385 | |
QXISTPDUYKNPLU-UHFFFAOYSA-N | |
2-bromo-1,4-dimethylbenzene | |
11121 | |
97% |
Chemical Identifiers
553-94-6 | |
185.06 | |
QXISTPDUYKNPLU-UHFFFAOYSA-N | |
11121 | |
CC1=CC=C(C)C(Br)=C1 |
C8H9Br | |
MFCD00000074 | |
2,5-dimethylbromobenzene, 2-bromo-p-xylene, 1-bromo-2,5-dimethylbenzene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, 2-bromo-1,4-dimethyl-benzene, 2,5-xylyl bromide, 2-bromo-1,4-xylene, 2,5-dimethylphenyl bromide, p-xylene, 2-bromo | |
2-bromo-1,4-dimethylbenzene |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
EINECSNumber : 209-055-7
TSCA : TSCA
RUO – Research Use Only