missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-p-xylene, 97%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 173421000

 View more versions of this product

Catalog No. AC173421000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Bromo-p-xylene
79°C
Glass bottle
1.34
Colorless
100g
553-94-6
100.0
C8H9Br
MFCD00000074
1-bromo-2,5-dimethylbenzene, 2,5-dimethylbromobenzene, 2,5-dimethylphenyl bromide, 2,5-xylyl bromide, 2-bromo-1,4-dimethyl-benzene, 2-bromo-1,4-xylene, 2-bromo-p-xylene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, p-xylene, 2-bromo
CC1=CC=C(C)C(Br)=C1
185.06
185.06
97%
1.3400g/mL
Authentic
1.5495 to 1.5515
199°C to 201°C
9°C to 10°C
97%
96.0
96% min. (GC)
(CH3)2C6H3Br
05, 385
QXISTPDUYKNPLU-UHFFFAOYSA-N
2-bromo-1,4-dimethylbenzene
11121
Liquid

Chemical Identifiers

553-94-6
185.06
QXISTPDUYKNPLU-UHFFFAOYSA-N
11121
CC1=CC=C(C)C(Br)=C1
C8H9Br
MFCD00000074
1-bromo-2,5-dimethylbenzene, 2,5-dimethylbromobenzene, 2,5-dimethylphenyl bromide, 2,5-xylyl bromide, 2-bromo-1,4-dimethyl-benzene, 2-bromo-1,4-xylene, 2-bromo-p-xylene, benzene, 2-bromo-1,4-dimethyl, bromo-p-xylene, p-xylene, 2-bromo
2-bromo-1,4-dimethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 209-055-7

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only