missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-N-(tert-butoxycarbonyl)benzylamine, TCI America™

$50.42 - $194.03

Chemical Identifiers

CAS 162356-90-3
Molecular Formula C12H16BrNO2
Molecular Weight (g/mol) 286.169
MDL Number MFCD05664385
InChI Key DFNZFCPEUDSNEO-UHFFFAOYSA-N
Synonym tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
PubChem CID 7018300
IUPAC Name tert-butyl N-[(2-bromophenyl)methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B49201G View Documents TCI America
B49201G
1g Each for $50.42
 
B49205G View Documents TCI America
B49205G
5g Each for $194.03
 
Specifications

Chemical Identifiers

162356-90-3
286.169
DFNZFCPEUDSNEO-UHFFFAOYSA-N
7018300
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
C12H16BrNO2
MFCD05664385
tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
tert-butyl N-[(2-bromophenyl)methyl]carbamate

Specifications

White
1g
C12H16BrNO2
tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
286.169
286.17
≥98.0% (GC)
53°C
162356-90-3
MFCD05664385
DFNZFCPEUDSNEO-UHFFFAOYSA-N
tert-butyl N-[(2-bromophenyl)methyl]carbamate
7018300
Crystal-Powder at 20°C
2-Bromo-N-(tert-butoxycarbonyl)benzylamine
Safety and Handling

Safety and Handling

TSCA : No