Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N,N-dimethylaniline, 98+%

$111.59 - $441.77

Chemical Identifiers

Chemical Name or Material 2-Bromo-N,N-dimethylaniline
CAS 698-00-0
Molecular Formula C8H10BrN
Formula Weight 200.08
MDL Number MFCD00013522
Beilstein 2205741
Synonym N,N-Dimethyl-2-bromoaniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2556406

  SDS
alfa aesar™
B2556406
5g Each for $111.59

AAB2556414

  SDS
alfa aesar™
B2556414
25g Each for $441.77
Description & Specifications

Specifications

Formula Weight 200.08
Synonym N,N-Dimethyl-2-bromoaniline
Chemical Name or Material 2-Bromo-N,N-dimethylaniline
Assay ≥98%
Molecular Formula C8H10BrN
MDL Number MFCD00013522
CAS 698-00-0
Beilstein 2205741
Boiling Point 108°C (14mmHg)
Quantity 5g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.