missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N,N-diisobutylbenzenesulfonamide, 97%

Chemical Identifiers

CAS 1308074-49-8
Molecular Formula C14H22BrNO2S
Molecular Weight (g/mol) 348.299
MDL Number MFCD13225051
InChI Key QYAKCDYMLXIWKK-UHFFFAOYSA-N
Synonym 2-bromo-n,n-bis 2-methylpropyl benzenesulfonamide, 2-bromo-n,n-diisobutylbenzenesulfonamide
PubChem CID 60707653
IUPAC Name 2-bromo-N,N-bis(2-methylpropyl)benzenesulfonamide
SMILES CC(C)CN(CC(C)C)S(=O)(=O)C1=CC=CC=C1Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60689MD View Documents Alfa Aesar™
H60689MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6068903 View Documents Alfa Aesar™
H6068903
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1308074-49-8
348.299
QYAKCDYMLXIWKK-UHFFFAOYSA-N
60707653
CC(C)CN(CC(C)C)S(=O)(=O)C1=CC=CC=C1Br
C14H22BrNO2S
MFCD13225051
2-bromo-n,n-bis 2-methylpropyl benzenesulfonamide, 2-bromo-n,n-diisobutylbenzenesulfonamide
2-bromo-N,N-bis(2-methylpropyl)benzenesulfonamide

Specifications

250mg
C14H22BrNO2S
2-bromo-n,n-bis 2-methylpropyl benzenesulfonamide, 2-bromo-n,n-diisobutylbenzenesulfonamide
CC(C)CN(CC(C)C)S(=O)(=O)C1=CC=CC=C1Br
348.299
348.3
2-Bromo-N,N-diisobutylbenzenesulfonamide
1308074-49-8
MFCD13225051
QYAKCDYMLXIWKK-UHFFFAOYSA-N
2-bromo-N,N-bis(2-methylpropyl)benzenesulfonamide
60707653
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning