missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N-methylbenzylamine, 95%

Manufacturer:  Alfa Aesar™ H6339003

 View more versions of this product

Catalog No. AAH6339003



Specifications

Specifications

2-Bromo-N-methylbenzylamine
1.5670
71°C to 74°C (2mmHg)
698-19-1
MFCD05262953
TUADRPBKJHMHDH-UHFFFAOYSA-N
1-(2-bromophenyl)-N-methylmethanamine
522360
Viscous Liquid
1.358g/mL
Air Sensitive
1g
C8H10BrN
2-bromobenzyl methylamine, n-methyl-2-bromobenzylamine, 2-bromo-n-methylbenzylamine, 1-2-bromophenyl-n-methylmethanamine, 2-bromophenyl methyl methyl amine, benzenemethanamine, 2-bromo-n-methyl, n-methyl-o-bromobenzylamine, acmc-20ao1g, 2-bromo-n-methyl benzylamine, 2-bromo-benzyl-methyl-amine
CNCC1=CC=CC=C1Br
200.079
200.08
95%

Chemical Identifiers

698-19-1
200.079
TUADRPBKJHMHDH-UHFFFAOYSA-N
522360
CNCC1=CC=CC=C1Br
C8H10BrN
MFCD05262953
2-bromobenzyl methylamine, n-methyl-2-bromobenzylamine, 2-bromo-n-methylbenzylamine, 1-2-bromophenyl-n-methylmethanamine, 2-bromophenyl methyl methyl amine, benzenemethanamine, 2-bromo-n-methyl, n-methyl-o-bromobenzylamine, acmc-20ao1g, 2-bromo-n-methyl benzylamine, 2-bromo-benzyl-methyl-amine
1-(2-bromophenyl)-N-methylmethanamine
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

storageNote1 : 2° to 8°C

SDS Documents