Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N-methylbenzylamine, 95%

Chemical Identifiers

Chemical Name or Material 2-Bromo-N-methylbenzylamine
CAS 698-19-1
Formula Weight 200.08
MDL Number MFCD05262953
Synonym (2-Bromobenzyl)methylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6339003

  SDS
alfa aesar™
H6339003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6339006

  SDS
alfa aesar™
H6339006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 71° to 74°C (2mmHg)
Formula Weight 200.08
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 95%
Sensitivity Air Sensitive
Chemical Name or Material 2-Bromo-N-methylbenzylamine
CAS 698-19-1
Synonym (2-Bromobenzyl)methylamine
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Refractive Index 1.5670
Health Hazard 1 Danger
MDL Number MFCD05262953
Storage Note 1 2° to 8°C
Physical Form Viscous Liquid
Density 1.358g/mL
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C