Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N-methylbenzamide, 99%

$192.86 - $840.77

Chemical Identifiers

Chemical Name or Material 2-Bromo-N-methylbenzamide
CAS 61436-88-2
Molecular Formula C8H8BrNO
Formula Weight 214.06
MDL Number MFCD00099250
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5058303

  SDS
alfa aesar™
H5058303
1g Each for $192.86

AAH5058306

  SDS
alfa aesar™
H5058306
5g Each for $840.77
Description & Specifications

Specifications

Formula Weight 214.06
MDL Number MFCD00099250
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2-Bromo-N-methylbenzamide
Health Hazard 1 Warning
Quantity 1g
Assay 99%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C8H8BrNO
CAS 61436-88-2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.