missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-N-methylaniline, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5516606
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-N-methylaniline | |
Brown | |
107°C to 109°C (12 mmHg) | |
C7H8BrN | |
MFCD06798064 | |
2-bromo-n-methylbenzenamine, acmc-20abqa, o-bromo-n-methylaniline, n-methyl-2-bromoaniline, benzenamine,2-bromo-n-methyl, 2-bromo-n-methylaniline, benzenamine, 2-bromo-n-methyl | |
CNC1=CC=CC=C1Br | |
186.052 | |
186.05 | |
Liquid |
6832-87-7 | |
1.589 g/mL | |
103°C (217°F) | |
1.607 | |
5 g | |
SMVIAQFTVWDWDS-UHFFFAOYSA-N | |
2-bromo-N-methylaniline | |
334086 | |
95% |
Chemical Identifiers
6832-87-7 | |
186.052 | |
SMVIAQFTVWDWDS-UHFFFAOYSA-N | |
2-bromo-N-methylaniline |
C7H8BrN | |
MFCD06798064 | |
334086 | |
CNC1=CC=CC=C1Br |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only