missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 1179183-69-7
Molecular Formula C14H14BrNO4S
Molecular Weight (g/mol) 372.233
MDL Number MFCD15526207
InChI Key WPZXLTFEOQPBPJ-UHFFFAOYSA-N
Synonym 2-bromo-n-3,4-dimethoxyphenyl benzenesulfonamide, 2-bromo-n-3,4-dimethoxy-phenyl-benzenesulfonamid
PubChem CID 3872247
IUPAC Name 2-bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide
SMILES COC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=CC=C2Br)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60073MD View Documents Alfa Aesar™
H60073MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6007303 View Documents Alfa Aesar™
H6007303
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1179183-69-7
372.233
WPZXLTFEOQPBPJ-UHFFFAOYSA-N
3872247
COC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=CC=C2Br)OC
C14H14BrNO4S
MFCD15526207
2-bromo-n-3,4-dimethoxyphenyl benzenesulfonamide, 2-bromo-n-3,4-dimethoxy-phenyl-benzenesulfonamid
2-bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide

Specifications

250mg
C14H14BrNO4S
2-bromo-n-3,4-dimethoxyphenyl benzenesulfonamide, 2-bromo-n-3,4-dimethoxy-phenyl-benzenesulfonamid
COC1=C(C=C(C=C1)NS(=O)(=O)C2=CC=CC=C2Br)OC
372.233
372.23
2-Bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide
1179183-69-7
MFCD15526207
WPZXLTFEOQPBPJ-UHFFFAOYSA-N
2-bromo-N-(3,4-dimethoxyphenyl)benzenesulfonamide
3872247
97%