missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N-(3,4-dichlorobenzyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 1421262-30-7
Molecular Formula C13H10BrCl2NO2S
Molecular Weight (g/mol) 395.092
MDL Number MFCD22683125
InChI Key BBKPFBULBLAWNE-UHFFFAOYSA-N
Synonym 2-bromo-n-3,4-dichlorobenzyl benzenesulfonamide, 2-bromo-n-3,4-dichlorophenyl methyl benzenesulfonamide
PubChem CID 97030408
IUPAC Name 2-bromo-N-[(3,4-dichlorophenyl)methyl]benzenesulfonamide
SMILES C1=CC=C(C(=C1)S(=O)(=O)NCC2=CC(=C(C=C2)Cl)Cl)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60420MD
View Documents
Alfa Aesar™
H60420MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6042003
View Documents
Alfa Aesar™
H6042003
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

1421262-30-7
395.092
BBKPFBULBLAWNE-UHFFFAOYSA-N
97030408
C1=CC=C(C(=C1)S(=O)(=O)NCC2=CC(=C(C=C2)Cl)Cl)Br
C13H10BrCl2NO2S
MFCD22683125
2-bromo-n-3,4-dichlorobenzyl benzenesulfonamide, 2-bromo-n-3,4-dichlorophenyl methyl benzenesulfonamide
2-bromo-N-[(3,4-dichlorophenyl)methyl]benzenesulfonamide

Specifications

250mg
C13H10BrCl2NO2S
2-bromo-n-3,4-dichlorobenzyl benzenesulfonamide, 2-bromo-n-3,4-dichlorophenyl methyl benzenesulfonamide
C1=CC=C(C(=C1)S(=O)(=O)NCC2=CC(=C(C=C2)Cl)Cl)Br
395.092
395.1
2-Bromo-N-(3,4-dichlorobenzyl)benzenesulfonamide
1421262-30-7
MFCD22683125
BBKPFBULBLAWNE-UHFFFAOYSA-N
2-bromo-N-[(3,4-dichlorophenyl)methyl]benzenesulfonamide
97030408
97%
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures