missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-N-(1-naphthyl)benzenesulfonamide, 97%

Chemical Identifiers

CAS 851215-65-1
Molecular Formula C16H12BrNO2S
Molecular Weight (g/mol) 362.241
MDL Number MFCD05617821
InChI Key LSOKWJIMCSVKBL-UHFFFAOYSA-N
Synonym 2-bromo-n-naphthalen-1-yl benzenesulfonamide, 2-bromo-n-1-naphthyl benzenesulfonamide, 2-bromo-n-naphthalen-1-yl benzene-1-sulfonamide
PubChem CID 2465063
IUPAC Name 2-bromo-N-naphthalen-1-ylbenzenesulfonamide
SMILES C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH60769MD View Documents Alfa Aesar™
H60769MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6076903 View Documents Alfa Aesar™
H6076903
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

851215-65-1
362.241
LSOKWJIMCSVKBL-UHFFFAOYSA-N
2465063
C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3Br
C16H12BrNO2S
MFCD05617821
2-bromo-n-naphthalen-1-yl benzenesulfonamide, 2-bromo-n-1-naphthyl benzenesulfonamide, 2-bromo-n-naphthalen-1-yl benzene-1-sulfonamide
2-bromo-N-naphthalen-1-ylbenzenesulfonamide

Specifications

250mg
C16H12BrNO2S
2-bromo-n-naphthalen-1-yl benzenesulfonamide, 2-bromo-n-1-naphthyl benzenesulfonamide, 2-bromo-n-naphthalen-1-yl benzene-1-sulfonamide
C1=CC=C2C(=C1)C=CC=C2NS(=O)(=O)C3=CC=CC=C3Br
362.241
362.24
2-Bromo-N-(1-naphthyl)benzenesulfonamide
851215-65-1
MFCD05617821
LSOKWJIMCSVKBL-UHFFFAOYSA-N
2-bromo-N-naphthalen-1-ylbenzenesulfonamide
2465063
97%