missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-D-phenylalanine, 95%

$70.11 - $830.68

Chemical Identifiers

CAS 267225-27-4
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD03092959
InChI Key JFVLNTLXEZDFHW-MRVPVSSYSA-N
Synonym 2-bromo-d-phenylalanine, 2r-2-amino-3-2-bromophenyl propanoic acid, d-2-br-phe-oh, d-2-bromophe, d-2-bromophenylalanine, h-d-phe 2-br-oh, o-bromo-d-phenylalanine, r-2-amino-3-2'-bromophenyl propanoic acid, r-2-amino-3-2-bromophenyl propanoic acid, r-2-bromophenylalanine
PubChem CID 7023575
IUPAC Name (2R)-2-amino-3-(2-bromophenyl)propanoic acid
SMILES C1=CC=C(C(=C1)CC(C(=O)O)N)Br
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63730MD
View Documents
Alfa Aesar™
H63730MD
250mg Each for $70.11
 
AAH6373003
View Documents
Alfa Aesar™
H6373003
1g Each for $208.49
 
AAH6373006
View Documents
Alfa Aesar™
H6373006
5g Each for $830.68
 
Specifications

Chemical Identifiers

267225-27-4
244.088
JFVLNTLXEZDFHW-MRVPVSSYSA-N
7023575
C1=CC=C(C(=C1)CC(C(=O)O)N)Br
C9H10BrNO2
MFCD03092959
2-bromo-d-phenylalanine, 2r-2-amino-3-2-bromophenyl propanoic acid, d-2-br-phe-oh, d-2-bromophe, d-2-bromophenylalanine, h-d-phe 2-br-oh, o-bromo-d-phenylalanine, r-2-amino-3-2'-bromophenyl propanoic acid, r-2-amino-3-2-bromophenyl propanoic acid, r-2-bromophenylalanine
(2R)-2-amino-3-(2-bromophenyl)propanoic acid

Specifications

267225-27-4
MFCD03092959
JFVLNTLXEZDFHW-MRVPVSSYSA-N
(2R)-2-amino-3-(2-bromophenyl)propanoic acid
7023575
Powder
White
2-Bromo-D-phenylalanine
C9H10BrNO2
2-bromo-d-phenylalanine, 2r-2-amino-3-2-bromophenyl propanoic acid, d-2-br-phe-oh, d-2-bromophe, d-2-bromophenylalanine, h-d-phe 2-br-oh, o-bromo-d-phenylalanine, r-2-amino-3-2'-bromophenyl propanoic acid, r-2-amino-3-2-bromophenyl propanoic acid, r-2-bromophenylalanine
C1=CC=C(C(=C1)CC(C(=O)O)N)Br
244.088
244.09
95%
250mg
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C