missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Bromo-D-phenylalanine, 95%

Manufacturer:  Alfa Aesar™ H6373003

 View more versions of this product

Catalog No. AAH6373003



Specifications

Specifications

2-Bromo-D-phenylalanine
C9H10BrNO2
2-bromo-d-phenylalanine, 2r-2-amino-3-2-bromophenyl propanoic acid, d-2-br-phe-oh, d-2-bromophe, d-2-bromophenylalanine, h-d-phe 2-br-oh, o-bromo-d-phenylalanine, r-2-amino-3-2'-bromophenyl propanoic acid, r-2-amino-3-2-bromophenyl propanoic acid, r-2-bromophenylalanine
C1=CC=C(C(=C1)CC(C(=O)O)N)Br
244.088
244.09
95%
1g
267225-27-4
MFCD03092959
JFVLNTLXEZDFHW-MRVPVSSYSA-N
(2R)-2-amino-3-(2-bromophenyl)propanoic acid
7023575
Powder
White

Chemical Identifiers

267225-27-4
244.088
JFVLNTLXEZDFHW-MRVPVSSYSA-N
7023575
C1=CC=C(C(=C1)CC(C(=O)O)N)Br
C9H10BrNO2
MFCD03092959
2-bromo-d-phenylalanine, 2r-2-amino-3-2-bromophenyl propanoic acid, d-2-br-phe-oh, d-2-bromophe, d-2-bromophenylalanine, h-d-phe 2-br-oh, o-bromo-d-phenylalanine, r-2-amino-3-2'-bromophenyl propanoic acid, r-2-amino-3-2-bromophenyl propanoic acid, r-2-bromophenylalanine
(2R)-2-amino-3-(2-bromophenyl)propanoic acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents